missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylenetetramine 60.0+%, TCI America™
$41.02 - $65.24
Chemical Identifiers
| CAS | 112-24-3 |
|---|---|
| Molecular Formula | C6H18N4 |
| Molecular Weight (g/mol) | 146.238 |
| MDL Number | MFCD00008169 |
| InChI Key | VILCJCGEZXAXTO-UHFFFAOYSA-N |
| Synonym | triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 |
| PubChem CID | 5565 |
| ChEBI | CHEBI:39501 |
| IUPAC Name | N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
| SMILES | C(CNCCNCCN)N |
Chemical Identifiers
| 112-24-3 | |
| 146.238 | |
| VILCJCGEZXAXTO-UHFFFAOYSA-N | |
| 5565 | |
| N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
| C6H18N4 | |
| MFCD00008169 | |
| triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
| CHEBI:39501 | |
| C(CNCCNCCN)N |
Specifications
| 112-24-3 | |
| 266°C | |
| MFCD00008169 | |
| 2259 | |
| VILCJCGEZXAXTO-UHFFFAOYSA-N | |
| N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine | |
| 5565 | |
| 146.24 | |
| Liquid |
| Yellow | |
| C6H18N4 | |
| 25 mL | |
| triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
| C(CNCCNCCN)N | |
| 146.238 | |
| CHEBI:39501 | |
| ≥60.0% (GC) | |
| Triethylenetetramine |
Safety and Handling
EINECSNumber : (2)-0163
RTECSNumber : YE6650000
TSCA : Yes