missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine 60.0+%, TCI America™

Catalog No. T0429500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0429500ML 500 mL
T042925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0429500ML Supplier TCI America Supplier No. T0429500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
MDL Number MFCD00008169
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N

Specifications

Color Yellow
Boiling Point 266°C
Quantity 500 mL
UN Number 2259
Formula Weight 146.24
Percent Purity ≥60.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylenetetramine
EINECSNumber (2)-0163
RTECSNumber YE6650000
TSCA Yes