missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol 99.0+%, TCI America™

Catalog No. p-7136276
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T042825G 25 g
T0428500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T042825G Supplier TCI America Supplier No. T042825G
Only null left

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
MDL Number MFCD00002880,MFCD00081839,MFCD01779596,MFCD01779599,MFCD01779601,MFCD01779603,MFCD01779605,MFCD01779609,MFCD01779611,MFCD01779612,MFCD01779614,MFCD01779615,MFCD01779616
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
Synonym Triglycol
PubChem CID 8172
ChEBI CHEBI:44926
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCCO

Specifications

Color Yellow
Physical Form Clear Liquid at 20°C
Boiling Point 276°C
Quantity 25 g
Formula Weight 150.17
Percent Purity ≥99.0% (GC)
Chemical Name or Material Triethylene Glycol
EINECSNumber (2)-0429
RTECSNumber YE4550000
TSCA Yes