missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol, Purified, 97%, Spectrum™ Chemical

Catalog No. 18614553
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614553 200 L Drum
18614552 500 mL Amber Glass Bottle
18614554 4 L Amber Glass Bottle
18614551 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614553 Supplier Spectrum Chemical Mfg Cor Supplier No. T1118200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCCO

Specifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 0.97
Grade Purified

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.