missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol, Purified, 97%, Spectrum™ Chemical

Catalog No. 18614552
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614552 500 mL Steel Can
18614554 4 L Steel Can
18614551 20 L Steel Can
18614553 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614552 Supplier Spectrum Chemical Mfg Cor Supplier No. T1118500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 112-27-6
Molecular Formula C6H14O4
Molecular Weight (g/mol) 150.17
InChI Key ZIBGPFATKBEMQZ-UHFFFAOYSA-N
IUPAC Name 2-[2-(2-hydroxyethoxy)ethoxy]ethan-1-ol
SMILES OCCOCCOCCO

Specifications

CAS Max % 1
Boiling Point 285°C to 287.4°C (545°F to 549.32°F)
Packaging Steel Can
Quantity 500 mL
Percent Purity 0.97
Grade Purified
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethylene Glycol

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.