missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Monoethyl Ether 90.0+%, TCI America™

Catalog No. E016725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E016725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E016725ML Supplier TCI America Supplier No. E016725ML

Chemical Identifiers

CAS 112-50-5
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD00020605
InChI Key WFSMVVDJSNMRAR-UHFFFAOYSA-N
PubChem CID 8190
IUPAC Name 2-[2-(2-ethoxyethoxy)ethoxy]ethan-1-ol
SMILES CCOCCOCCOCCO

Specifications

Melting Point -19°C
Color Colorless
Boiling Point 255°C
Quantity 25 mL
Formula Weight 178.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Monoethyl Ether
EINECSNumber (2)-0436&(7)-0097
RTECSNumber KK8950000
TSCA Yes