missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Monobutyl Ether 98.0+%, TCI America™
$52.19 - $100.67
Chemical Identifiers
| CAS | 143-22-6 |
|---|---|
| Molecular Formula | C10H22O4 |
| Molecular Weight (g/mol) | 206.28 |
| MDL Number | MFCD00020606 |
| InChI Key | COBPKKZHLDDMTB-UHFFFAOYSA-N |
| Synonym | triethylene glycol monobutyl ether, butoxytriglycol, dowanol tbat, butoxytriethylene glycol, poly-solv tb, triethylene glycol butyl ether, triglycol monobutyl ether, ethanol, 2-2-2-butoxyethoxy ethoxy, triethylene glycol mono-n-butyl ether, triethylene glycol n-butyl ether |
| PubChem CID | 8923 |
| IUPAC Name | 2-[2-(2-butoxyethoxy)ethoxy]ethan-1-ol |
| SMILES | CCCCOCCOCCOCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T091525ML
|
TCI America
T091525ML |
25 mL |
Each for $52.19
|
|
|||||
|
T0915500ML
|
TCI America
T0915500ML |
500 mL |
Each for $100.67
|
|
|||||
Chemical Identifiers
| 143-22-6 | |
| 206.28 | |
| COBPKKZHLDDMTB-UHFFFAOYSA-N | |
| 8923 | |
| CCCCOCCOCCOCCO |
| C10H22O4 | |
| MFCD00020606 | |
| triethylene glycol monobutyl ether, butoxytriglycol, dowanol tbat, butoxytriethylene glycol, poly-solv tb, triethylene glycol butyl ether, triglycol monobutyl ether, ethanol, 2-2-2-butoxyethoxy ethoxy, triethylene glycol mono-n-butyl ether, triethylene glycol n-butyl ether | |
| 2-[2-(2-butoxyethoxy)ethoxy]ethan-1-ol |
Specifications
| 143-22-6 | |
| Yellow | |
| C10H22O4 | |
| 25 mL | |
| COBPKKZHLDDMTB-UHFFFAOYSA-N | |
| 2-[2-(2-butoxyethoxy)ethoxy]ethan-1-ol | |
| 8923 | |
| ≥98.0% (GC) | |
| Triethylene Glycol Monobutyl Ether |
| -35°C | |
| 278°C | |
| MFCD00020606 | |
| triethylene glycol monobutyl ether, butoxytriglycol, dowanol tbat, butoxytriethylene glycol, poly-solv tb, triethylene glycol butyl ether, triglycol monobutyl ether, ethanol, 2-2-2-butoxyethoxy ethoxy, triethylene glycol mono-n-butyl ether, triethylene glycol n-butyl ether | |
| CCCCOCCOCCOCCO | |
| 206.28 | |
| 206.28 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0436&(7)-0097
RTECSNumber : KJ9450000
TSCA : Yes