missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Divinyl Ether (stabilized with KOH) 98.0+%, TCI America™

Catalog No. T346025G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T346025G 25 g
T3460100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T346025G Supplier TCI America Supplier No. T346025G
Only null left

Chemical Identifiers

CAS 765-12-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00074891
InChI Key CYIGRWUIQAVBFG-UHFFFAOYSA-N
PubChem CID 3033992
IUPAC Name 1,2-bis(2-ethenoxyethoxy)ethane
SMILES C=COCCOCCOCCOC=C

Specifications

Color Colorless
Boiling Point 126°C
Quantity 25 g
Formula Weight 202.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Divinyl Ether (stabilized with KOH)
EINECSNumber (7)-1321
RTECSNumber XF0877100
TSCA Yes