missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol Butyl Methyl Ether 98.0+%, TCI America™
$37.29 - $98.79
Chemical Identifiers
| CAS | 7382-30-1 |
|---|---|
| Molecular Formula | C11H24O4 |
| Molecular Weight (g/mol) | 220.31 |
| MDL Number | MFCD23703159 |
| InChI Key | SNAQINZKMQFYFV-UHFFFAOYSA-N |
| Synonym | 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane |
| PubChem CID | 18344639 |
| IUPAC Name | 2,5,8,11-tetraoxapentadecane |
| SMILES | CCCCOCCOCCOCCOC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T318125ML
|
TCI America
T318125ML |
25 mL |
Each for $37.29
|
|
|||||
|
T3181500ML
|
TCI America
T3181500ML |
500 mL |
Each for $98.79
|
|
|||||
Chemical Identifiers
| 7382-30-1 | |
| 220.31 | |
| SNAQINZKMQFYFV-UHFFFAOYSA-N | |
| 18344639 | |
| CCCCOCCOCCOCCOC |
| C11H24O4 | |
| MFCD23703159 | |
| 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane | |
| 2,5,8,11-tetraoxapentadecane |
Specifications
| 7382-30-1 | |
| Colorless | |
| MFCD23703159 | |
| 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane | |
| CCCCOCCOCCOCCOC | |
| 220.31 | |
| 220.31 | |
| Liquid |
| -34°C | |
| C11H24O4 | |
| 25 mL | |
| SNAQINZKMQFYFV-UHFFFAOYSA-N | |
| 2,5,8,11-tetraoxapentadecane | |
| 18344639 | |
| ≥98.0% (GC) | |
| Triethylene Glycol Butyl Methyl Ether |
Safety and Handling
EINECSNumber : (7)-1321
TSCA : No