missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylene Glycol 2-Bromoethyl Methyl Ether 95.0+%, TCI America™
$331.81 - $986.08
Chemical Identifiers
| CAS | 110429-45-3 |
|---|---|
| Molecular Formula | C9H19BrO4 |
| Molecular Weight (g/mol) | 271.151 |
| InChI Key | YFFFQGXWMHAJPP-UHFFFAOYSA-N |
| Synonym | 2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide |
| PubChem CID | 19389132 |
| IUPAC Name | 1-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]-2-methoxyethane |
| SMILES | COCCOCCOCCOCCBr |
Chemical Identifiers
| 110429-45-3 | |
| 271.151 | |
| 2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide | |
| 1-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]-2-methoxyethane |
| C9H19BrO4 | |
| YFFFQGXWMHAJPP-UHFFFAOYSA-N | |
| 19389132 | |
| COCCOCCOCCOCCBr |
Specifications
| 110429-45-3 | |
| C9H19BrO4 | |
| 2-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]ethyl Bromide, 1-Bromo-3,6,9,12-tetraoxatridecane, mPEG4-Bromide | |
| COCCOCCOCCOCCBr | |
| 271.151 | |
| 271.15 | |
| Liquid |
| Yellow | |
| 5 g | |
| YFFFQGXWMHAJPP-UHFFFAOYSA-N | |
| 1-[2-[2-(2-bromoethoxy)ethoxy]ethoxy]-2-methoxyethane | |
| 19389132 | |
| ≥95.0% (GC) | |
| Triethylene Glycol 2-Bromoethyl Methyl Ether |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator