missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Fluka™ Triethylammonium Bicarbonate Buffer, Honeywell™ Fluka™

Catalog No. 60044973
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60044973 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60044973 Supplier Honeywell Fluka™ Supplier No. 17902100ML
Only null left

volatile buffer, for HPLC, 1 M

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications

CAS 15715-58-9
Color Colorless
For Use With (Equipment) Volatile buffer for HPLC
pH 8.4 to 8.6 (at 25°C)
Physical Form Liquid
Quantity 100 mL
Packaging Glass Bottle
Molecular Weight (g/mol) 163.217