Learn More
MilliporeSigma™ Triethylammonium Acetate, 1 M Solution, Calbiochem™,
Catalog No. 6257181l
Change view
Click to view available options
Quantity:
1L
Catalog No. Quantity
6257181L 1L
1 options
Catalog No. 6257181L Supplier MilliporeSigma™ Supplier No. 6257181L
Only null left
Ready-to-use buffer solution used for the purification of chemically synthesized oligonucleotides by HPLC
Specifications
CAS 5204-74-0
Quantity 1L