missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine 99.0+%, TCI America™

Catalog No. T0424500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0424500ML 500 mL
T042425ML 25 mL
T0424100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0424500ML Supplier TCI America Supplier No. T0424500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115°C
Color Colorless
Boiling Point 89°C
Quantity 500 mL
UN Number 1296
Formula Weight 101.19
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Triethylamine
EINECSNumber (2)-0141
RTECSNumber YE0175000
TSCA Yes