missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine (Reagent), Fisher Chemical™

Catalog No. O48854
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O48854 4 L Glass Bottle
O48851 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O48854 Supplier Thermo Fisher Scientific Supplier No. O4885 4
Only null left

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115°C
Color Colorless
Density 0.728g/cm³
pH 12.4
Boiling Point 90°C
Assay Percent Range ≥99%
Packaging Glass Bottle
Linear Formula (C2H5)3N
Quantity 4 L
UN Number 1296
Vapor Pressure 69mbar at 20°C
Formula Weight 101.19
Percent Purity ≥99%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethylamine
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Highly flammable. Harmful by inhalation, in contact with skin and if swallowed. May cause pulmonary edema. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid