missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylamine Hydrochloride 98.5+%, TCI Americaâ„¢
$54.81 - $202.21
Chemical Identifiers
CAS | 554-68-7 |
---|---|
Molecular Formula | C6H16ClN |
Molecular Weight (g/mol) | 137.65 |
MDL Number | MFCD00012500 |
InChI Key | ILWRPSCZWQJDMK-UHFFFAOYSA-N |
Synonym | triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochlorideShow More |
PubChem CID | 11130 |
IUPAC Name | hydrogen triethylamine chloride |
SMILES | [H+].[Cl-].CCN(CC)CC |
Chemical Identifiers
554-68-7 | |
137.65 | |
ILWRPSCZWQJDMK-UHFFFAOYSA-N | |
11130 | |
[H+].[Cl-].CCN(CC)CC |
C6H16ClN | |
MFCD00012500 | |
triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochlorideShow More | |
hydrogen triethylamine chloride |
Specifications
554-68-7 | |
White | |
MFCD00012500 | |
triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride | |
[H+].[Cl-].CCN(CC)CC | |
137.65 | |
137.65 | |
Crystalline Powder |
258°C | |
C6H16ClN | |
25 g | |
ILWRPSCZWQJDMK-UHFFFAOYSA-N | |
hydrogen triethylamine chloride | |
11130 | |
≥98.5% (T) | |
Triethylamine Hydrochloride |
Safety and Handling
EINECSNumber : (2)-0141
RTECSNumber : YE2111500
TSCA : Yes