missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine Hydrochloride 98.5+%, TCI America™

Catalog No. T0425500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0425500G 500 g
T042525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0425500G Supplier TCI America Supplier No. T0425500G

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
Synonym triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride
PubChem CID 11130
IUPAC Name hydrogen triethylamine chloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point 258°C
Color White
Quantity 500 g
Formula Weight 137.65
Percent Purity ≥98.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylamine Hydrochloride
EINECSNumber (2)-0141
RTECSNumber YE2111500
TSCA Yes