missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine hydrochloride, 99+%

Catalog No. p-3737618
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270672500 250 g Glass bottle
AC270670010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC270672500 Supplier Thermo Scientific Chemicals Supplier No. 270672500

CAS: 554-68-7 | C6H16ClN | 137.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 554-68-7
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00012500
InChI Key ILWRPSCZWQJDMK-UHFFFAOYSA-N
Synonym triethylamine hydrochloride, triethylammonium chloride, triethylamine hcl, ethanamine, n,n-diethyl-, hydrochloride, triethylamine monohydrochloride, triethylamine, hydrochloride, unii-0nx3818gcw, amine, triethyl, hydrochloride, n,n-diethylethanamine hydrochloride, triethylaminehydrochloride
PubChem CID 11130
IUPAC Name N,N-diethylethanamine;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CC

Specifications

Melting Point 254.0°C to 260.0°C
Color White
pH 4.5 to 6.5 (1% w/v soln.)
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (C2H5)3N·HCl
Quantity 250 g
Beilstein 04, 101
Merck Index 14, 9666
Solubility Information (10% in water) Clear colorless
Formula Weight 137.66
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Triethylamine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-067-2

RUO – Research Use Only