missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine hydrobromide, 99%

CAS: 636-70-4 | C6H16BrN | 182.11 g/mol

$72.04 - $72.04

Chemical Identifiers

CAS 636-70-4
Molecular Formula C6H16BrN
Molecular Weight (g/mol) 182.11
MDL Number MFCD00054287
InChI Key NRTLTGGGUQIRRT-UHFFFAOYSA-N
Synonym triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
PubChem CID 2734074
SMILES [Br-].CC[NH+](CC)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC349721000
SDS View Documents
Thermo Scientific Chemicals
349721000
100 g Glass bottle
Each for $72.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

636-70-4
182.11
NRTLTGGGUQIRRT-UHFFFAOYSA-N
2734074
C6H16BrN
MFCD00054287
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
[Br-].CC[NH+](CC)CC

Specifications

636-70-4
>65°C
98.5% min. (Argentometry)
C6H16BrN
MFCD00054287
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
[Br-].CC[NH+](CC)CC
182.11
182.1
Crystalline Powder or Flakes
White
Authentic
Glass bottle
(C2H5)3N·HBr
100 g
NRTLTGGGUQIRRT-UHFFFAOYSA-N
triethylazanium bromide
2734074
99%
Triethylamine hydrobromide, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISO

GHS Signal Word: Warning

EINECSNumber : 211-263-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only