missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine (HPLC), Fisher Chemical™

Catalog No. p-8847003
Change view
Click to view available options
Quantity:
500 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O4884100 100 mL
O4884500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O4884100 Supplier Thermo Fisher Scientific Supplier No. O4884 100
Only null left

Improve HPLC separation of acidic analytes by ion pairing.

  • High purity
  • Application tested

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

Packaging Glass Bottle
Quantity 100 mL