missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine (HPLC), Fisher Chemical™

Catalog No. O4884500
Change view
Click to view available options
Quantity:
500 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O4884500 500 mL
O4884100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O4884500 Supplier Thermo Fisher Scientific Supplier No. O4884500
  • ONLINE EXCLUSIVE PRICE
    $314.61
    $233.90 / Each

    Save $80.71

    26.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Improve HPLC separation of acidic analytes by ion pairing.

  • High purity
  • Application tested

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
IUPAC Name triethylamine
SMILES CCN(CC)CC

Specifications

Quantity 500 mL