missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, HPLC, >99.5%, MilliporeSigma™

Catalog No. MTX12025
Change view
Click to view available options
Quantity:
686 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX12025 686 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX12025 Supplier Technidata Supplier No. TX12025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, triethyl amine, trietilamina, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026

Specifications

Assay Percent Range >99.5%
Linear Formula (CH3CH2)3N
Quantity 686 mL
Formula Weight 11.19
Grade HPLC
Physical Form Liquid
Recommended Storage Room Temperature