missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, MilliporeSigma™

Catalog No. MTX120011
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX120011 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MTX120011 Supplier Technidata Supplier No. TX120011

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121-44-8
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -7.2°C
Density 0.729g/cm3
Boiling Point 89.4°C
Flash Point Closed cup: -11°C (12.2°F); Open cup: -6.15°C (20.9°F)
Ignition Point 248.85°C (479.9°F)
Odor Ammoniacal
Quantity 100 mL
UN Number UN 1296
Solubility Information Partially soluble in water
Formula Weight 101.19
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Triethylamine
Show More Show Less
DOTInformation DOT Class: 3 (8), Packaging Group: II
Recommended Storage Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.