Learn More
Triethylamine, 99%
Description
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Triethylamine is a base used to prepare esters and amides from acyl chlorides as well as in the synthesis of quaternary ammonium compounds. It acts as a catalyst in the formation of urethane foams and epoxy resins, dehydrohalogeantion reactions, acid neutralizer for condensation reactions and Swern oxidations. It finds application in reverse phase high-performance liquid chromatography (HPLC) as a mobile-phase modifier. It is also used as an accelerator activator for rubber, as a propellant, as a corrosion inhibitor, as a curing and hardening agent for polymers and for the desalination of seawater. Furthermore, it is used in automotive casting industry and textile industry.
Solubility
Miscible with water, ether and ethanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -115°C |
| Density | 0.726 |
| Boiling Point | 89°C to 90°C |
| Flash Point | −11°C (12°F) |
| Odor | Ammonia-like |
| Assay Percent Range | 99% |
| Refractive Index | 1.4005 |
| Linear Formula | (CH3CH2)3N |
| Quantity | 250 mL |
| UN Number | UN1296 |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H311+H331-H314-H335-H336 |
| Hazard Statement | GHS H Statement H225-H314-H302-H312-H332 Highly flammable liquid and vapour. Causes severe skin burns and eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIETHYLAMINE |
| EINECSNumber | 204-469-4 |
| RTECSNumber | YE0175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only