Learn More
Triethylamine, 99%, pure
CAS: 121-44-8 | C6H15N | 101.19 g/mol
Supplier: Thermo Scientific Chemicals 157910010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Triethylamine | |
| -115.0°C | |
| 90.0°C | |
| Authentic | |
| EcoSafPak™ Glass Bottle | |
| 1.3990 to 1.4010 | |
| 1 L | |
| 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336 | |
| 15, 9831 | |
| Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride | |
| CCN(CC)CC | |
| 101.193 | |
| CHEBI:35026 | |
| 69mbar at 20°C | |
| Pure |
| 121-44-8 | |
| 0.7280g/mL | |
| −11°C | |
| 99% | |
| C6H15N | |
| (C2H5)3N | |
| 04, 99 | |
| 0.728 | |
| triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
| ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
| N,N-diethylethanamine | |
| 8471 | |
| 101.19 | |
| 99% | |
| Liquid |
Chemical Identifiers
| 121-44-8 | |
| triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane | |
| CHEBI:35026 | |
| CCN(CC)CC |
| ZMANZCXQSJIPKH-UHFFFAOYSA-N | |
| 8471 | |
| N,N-diethylethanamine |
Safety and Handling
GHS P Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse
Danger
RUO – Research Use Only