missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99.7%, extra pure

CAS: 121-44-8 | C6H15N | 101.19 g/mol

Supplier:  Thermo Scientific Chemicals 219510025

 View more versions of this product

Catalog No. AC219510025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triethylamine
121-44-8
0.7280g/mL
−11°C
99.6% min. (GC)
C6H15N
(C2H5)3N
2.5 kg
01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
15, 9831
Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
CCN(CC)CC
101.19
0.36 mPa.s (20°C)
101.19
99.7%
Liquid
99.7%
-115.0°C
90.0°C
Authentic
Plastic Bottle
1.3990 to 1.4010
MFCD00009051
04, 99
0.728
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
ZMANZCXQSJIPKH-UHFFFAOYSA-N
N,N-diethylethanamine
8471
CHEBI:35026
69mbar at 20°C
Extra Pure

Chemical Identifiers

121-44-8
ZMANZCXQSJIPKH-UHFFFAOYSA-N
8471
N,N-diethylethanamine
MFCD00009051
triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
CHEBI:35026
CCN(CC)CC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 204-469-4

RTECSNumber : YE0175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only