missing translation for 'onlineSavingsMsg'
Learn More

Triethylamine, 99.7%, extra pure

Catalog No. AC219510500
Change view
Click to view available options
Quantity:
5 g
50 g
1 kg
2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC219510500 50 g
AC219510050 5 g
AC219510010 1 kg
AC219510025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC219510500 Supplier Thermo Scientific Chemicals Supplier No. 219510500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-44-8 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-44-8
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 99.7%
Molecular Formula C6H15N
Refractive Index 1.3990 to 1.4010
Linear Formula (C2H5)3N
Quantity 50 g
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Specific Gravity 0.728
Merck Index 15, 9831
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.193
Vapor Pressure 69mbar at 20°C
Formula Weight 101.19
Percent Purity 99.7%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Triethylamine, 99.7%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement Highly flammable liquid and vapour. Toxic if inhaled. Toxic in contact with skin. Harmful if swallowed. Causes severe skin burns and eye damage. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse
EINECSNumber 204-469-4
RTECSNumber YE0175000
TSCA TSCA

RUO – Research Use Only