missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Triethylamine

Catalog No. 4712834L
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4712834L 4 L
471283100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4712834L Supplier Sigma Aldrich Supplier No. 4712834L

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylamine

Specifications

CAS 121-44-8
Melting Point -115°C
Density 0.726 g/mL at 25°C
Boiling Point 88.8°C
Molecular Formula C6H15N
Refractive Index n20/D 1.401
Linear Formula (C2H5)3N
MDL Number MFCD00009051
Quantity 4 L
Synonym N,N-Diethylethanamine
Molecular Weight (g/mol) 101.19 g/mol
Show More Show Less