missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Triethylamine

Catalog No. 9034025L
Encompass
Change view
Click to view available options
Quantity:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9034025L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9034025L Supplier Sigma Aldrich Supplier No. 903402.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylamine

Specifications

CAS 121-44-8
Melting Point -115°C
Density 0.727 g/mL at 20°C, 0.726 g/mL at 25°C
Boiling Point 88.8°C
Molecular Formula C6H15N
Refractive Index n20/D 1.401; n20/D 1.401
Linear Formula (C2H5)3N
MDL Number MFCD00009051
Quantity 2.5 L
Synonym N,N-Diethylethanamine
Molecular Weight (g/mol) 101.19 g/mol
Show More Show Less