missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl(trifluoromethyl)silane 98.0+%, TCI America™
$117.43 - $352.30
Chemical Identifiers
| CAS | 120120-26-5 |
|---|---|
| Molecular Formula | C7H15F3Si |
| Molecular Weight (g/mol) | 184.28 |
| MDL Number | MFCD00192520 |
| InChI Key | ZHSKFONQCREGOG-UHFFFAOYSA-N |
| Synonym | (Triethylsilyl)trifluoromethane |
| PubChem CID | 4204914 |
| IUPAC Name | triethyl(trifluoromethyl)silane |
| SMILES | CC[Si](CC)(CC)C(F)(F)F |
Chemical Identifiers
| 120120-26-5 | |
| 184.28 | |
| ZHSKFONQCREGOG-UHFFFAOYSA-N | |
| 4204914 | |
| CC[Si](CC)(CC)C(F)(F)F |
| C7H15F3Si | |
| MFCD00192520 | |
| (Triethylsilyl)trifluoromethane | |
| triethyl(trifluoromethyl)silane |
Specifications
| 120120-26-5 | |
| 58°C | |
| MFCD00192520 | |
| 1993 | |
| ZHSKFONQCREGOG-UHFFFAOYSA-N | |
| triethyl(trifluoromethyl)silane | |
| 4204914 | |
| ≥98.0% (GC) | |
| Triethyl(trifluoromethyl)silane |
| Yellow | |
| C7H15F3Si | |
| 1 g | |
| (Triethylsilyl)trifluoromethane | |
| CC[Si](CC)(CC)C(F)(F)F | |
| 184.28 | |
| 184.28 | |
| Liquid |
Safety and Handling
TSCA : No