missing translation for 'onlineSavingsMsg'
Learn More

Triethyl(trifluoromethyl)silane 98.0+%, TCI America™

Catalog No. T27641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T27641G 1 g
T27645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T27641G Supplier TCI America Supplier No. T27641G
Only null left

Chemical Identifiers

CAS 120120-26-5
Molecular Formula C7H15F3Si
Molecular Weight (g/mol) 184.28
MDL Number MFCD00192520
InChI Key ZHSKFONQCREGOG-UHFFFAOYSA-N
Synonym (Triethylsilyl)trifluoromethane
PubChem CID 4204914
IUPAC Name triethyl(trifluoromethyl)silane
SMILES CC[Si](CC)(CC)C(F)(F)F

Specifications

Color Yellow
Boiling Point 58°C
Quantity 1 g
UN Number 1993
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl(trifluoromethyl)silane