missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Phosphonoacetate 97.0+%, TCI America™

Catalog No. p-7126765
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
D152325G 25 g
D1523100G 100 g
D1523500G 500 g
3 options

Catalog No. D152325G

Supplier: TCI America D152325G

Only null left

Chemical Identifiers

CAS 867-13-0
Molecular Formula C8H17O5P
Molecular Weight (g/mol) 224.193
MDL Number MFCD00009177
InChI Key GGUBFICZYGKNTD-UHFFFAOYSA-N
Synonym triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
PubChem CID 13345
IUPAC Name ethyl 2-diethoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OCC)OCC

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
UN Number 3082
Formula Weight 224.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Phosphonoacetate
EINECSNumber (2)-1961
RTECSNumber AG9800000
TSCA Yes