missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphonoacetate, 97%

Catalog No. p-4509978
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139701000 100 g Glass bottle
AC139705000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC139701000 Supplier Thermo Scientific Chemicals Supplier No. 139701000
Only null left

CAS: 867-13-0 | C8H17O5P | 224.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 867-13-0
Molecular Formula C8H17O5P
Molecular Weight (g/mol) 224.19
InChI Key GGUBFICZYGKNTD-UHFFFAOYSA-N
Synonym triethyl phosphonoacetate, ethyl 2-diethoxyphosphoryl acetate, ethyl diethoxyphosphoryl acetate, ethyl diethylphosphono acetate, triethyl phosphonoethanoate, diethyl carboethoxymethylphosphonate, diethyl ethoxycarbonylmethanephosphonate, diethylphosphonoacetic acid ethyl ester, diethyl carbethoxymethylphosphonate
PubChem CID 13345
IUPAC Name ethyl 2-diethoxyphosphorylacetate
SMILES CCOC(=O)CP(=O)(OCC)OCC

Specifications

Acidity 2mg KOH/g max.
Melting Point -24.0°C
Density 1.1300g/mL
Boiling Point 260.0°C to 262.0°C
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4300 to 1.4320
Linear Formula (C2H5O)2P(O)CH2CO2C2H5
Quantity 100 g
Beilstein 04,I,573
Fieser 01,1216; 06,612; 12,527; 13,255
Specific Gravity 1.13
Solubility Information Solubility in water: insoluble
Formula Weight 224.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Triethyl phosphonoacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 212-757-6

RUO – Research Use Only