missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Triethyl phosphate

Catalog No. 538728100ML
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
538728100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 538728100ML Supplier Sigma Aldrich Supplier No. 538728100ML

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Triethyl phosphate is an alkyl phosphate
  • It participates in the thermal transformation of sodium acetate-2-c14 to n-butyl acetate-2-c14.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 538728.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin.reagentplus is a registered trademark of sigma-aldrich co
  • Llc

Specifications

CAS 78-40-0
Density 1.072 g/mL (at 25°C (literature))
Boiling Point 215°C (lit.)
Molecular Formula C6H15O4P
Refractive Index n20/D 1.403 (literature)
Linear Formula (C2H5O)3 PO
MDL Number MFCD00009077
Quantity 100 mL
Synonym TEP; Ethyl phosphate
Molecular Weight (g/mol) 182.15
Percent Purity ≥99.8%
Show More Show Less