missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthovalerate 95.0+%, TCI America™

Catalog No. T097725ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097725ML 25 mL
T0977250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T097725ML Supplier TCI America Supplier No. T097725ML
Only null left

Chemical Identifiers

CAS 919-29-9
Molecular Formula C11H24O3
Molecular Weight (g/mol) 204.31
MDL Number MFCD00059383
InChI Key DIKAUBKIDNXNNW-UHFFFAOYSA-N
Synonym 1,1,1-Triethoxypentane, Orthovaleric Acid Triethyl Ester
PubChem CID 70195
IUPAC Name 1,1,1-triethoxypentane
SMILES CCCCC(OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 50°C
Quantity 25 mL
UN Number 3272
Formula Weight 204.31
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthovalerate