missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthoformate 98.0+%, TCI America™

Catalog No. O006625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O006625ML 25 mL
O0066500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O006625ML Supplier TCI America Supplier No. O006625ML

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -76°C
Color Yellow
Boiling Point 145°C
Quantity 25 mL
UN Number 2524
Formula Weight 148.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthoformate
EINECSNumber (2)-0683
RTECSNumber RM6475000
TSCA Yes