Learn More
Triethyl orthoformate, 98%
CAS: 122-51-0 | C7H16O3 | 148.202 g/mol
Supplier: Thermo Scientific Chemicals A135870D
Description
Triethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethyl orthoformate is used in Bodroux-Chichibabin aldehyde synthesis to prepare an aldehyde with one carbon higher by reacting with Grignard reagent. It is also used in the electrophilic formylation of activated aromatic species such as phenol.
Solubility
Miscible with alcohols, acetone, ethers and chloroform. Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with acids and strong oxidizing agents.
Specifications
| Triethyl orthoformate | |
| -61°C | |
| 144°C to 146°C | |
| 98% | |
| 1.391 | |
| MFCD00009230 | |
| UN2524 | |
| Moisture sensitive | |
| triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
| GKASDNZWUGIAMG-UHFFFAOYSA-N | |
| diethoxymethoxyethane | |
| 31214 | |
| 98% |
| 122-51-0 | |
| 0.891 | |
| 30°C (86°F) | |
| C7H16O3 | |
| HC(OCH2CH3)3 | |
| 10,000 mL | |
| 605384 | |
| 14,6884 | |
| Miscible with alcohols,acetone,ethers and chloroform. Slightly miscible with water. | |
| CCOC(OCC)OCC | |
| 148.202 | |
| 148.2 | |
| Liquid |
Chemical Identifiers
| 122-51-0 | |
| 148.202 | |
| GKASDNZWUGIAMG-UHFFFAOYSA-N | |
| 31214 | |
| CCOC(OCC)OCC |
| C7H16O3 | |
| MFCD00009230 | |
| triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty | |
| diethoxymethoxyethane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYL ORTHOFORMATE
EINECSNumber : 204-550-4
RTECSNumber : RM6475000
TSCA : Yes
storageNote1 : Moisture sensitive
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only