missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoformate, 98%

Catalog No. p-3737634
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139661000 100 mL Glass bottle
AC139660010 1 L Glass bottle
AC139660025 2.5 L Glass bottle
AC139660250 25 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC139661000 Supplier Thermo Scientific Chemicals Supplier No. 139661000
Only null left

CAS: 122-51-0 | C7H16O3 | 148.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent

Chemical Identifiers

CAS 122-51-0
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.2
MDL Number MFCD00009230
InChI Key GKASDNZWUGIAMG-UHFFFAOYSA-N
Synonym triethyl orthoformate, triethoxymethane, ethyl orthoformate, aethon, ethone, diethoxymethoxy ethane, methane, triethoxy, ethyl formate ortho, orthoformic acid triethyl ester, orthomravencan ethylnaty
PubChem CID 31214
IUPAC Name diethoxymethoxyethane
SMILES CCOC(OCC)OCC

Specifications

Melting Point -61.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 146.0°C
Flash Point 35°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3900 to 1.3920
Linear Formula CH(OC2H5)3
Quantity 100 mL
Beilstein 02, 20
Fieser 01,1204; 02,430; 04,527; 05,690; 06,610; 07,385; 11,555; 13,327
Specific Gravity 0.89
Merck Index 15, 6980
Solubility Information Solubility in water: 1.35g/L (20°C). Other solubilities: decomposes in water,soluble in alcohols,acetone,ethers,chloroform,
Formula Weight 148.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl orthoformate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 204-550-4

RUO – Research Use Only