missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthoacetate 96.0+%, TCI America™

Catalog No. O010525ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O010525ML 25 mL
O0105500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O010525ML Supplier TCI America Supplier No. O010525ML

Chemical Identifiers

CAS 78-39-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009223
InChI Key NDQXKKFRNOPRDW-UHFFFAOYSA-N
Synonym triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
PubChem CID 66221
IUPAC Name 1,1,1-triethoxyethane
SMILES CCOC(C)(OCC)OCC

Specifications

Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 3272
Formula Weight 162.23
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthoacetate
EINECSNumber (2)-2544
TSCA Yes