Learn More
Triethyl orthoacetate, 97%
CAS: 78-39-7 | C8H18O3 | 162.229 g/mol
Supplier: Thermo Scientific Chemicals L060040F
Description
Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTriethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.
Solubility
Slightly miscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
| Triethyl orthoacetate | |
| 0.887 | |
| 32°C (89°F) | |
| 1.397 | |
| 2500 mL | |
| 506201 | |
| triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane | |
| NDQXKKFRNOPRDW-UHFFFAOYSA-N | |
| 1,1,1-triethoxyethane | |
| 66221 | |
| 98% |
| 78-39-7 | |
| 142°C to 143°C | |
| C8H18O3 | |
| MFCD00009223 | |
| UN1993 | |
| Moisture sensitive | |
| Slightly miscible with water. | |
| CCOC(C)(OCC)OCC | |
| 162.229 | |
| 162.23 |
Chemical Identifiers
| 78-39-7 | |
| 162.229 | |
| NDQXKKFRNOPRDW-UHFFFAOYSA-N | |
| 66221 | |
| CCOC(C)(OCC)OCC |
| C8H18O3 | |
| MFCD00009223 | |
| triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane | |
| 1,1,1-triethoxyethane |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 201-112-4
RTECSNumber : KJ4075000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only