missing translation for 'onlineSavingsMsg'
Learn More

Triethyl orthoacetate, 97%

Catalog No. AAL06004AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL06004AP 500 mL
AAL06004AE 100 mL
AAL060040F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL06004AP Supplier Thermo Scientific Chemicals Supplier No. L06004AP
Only null left

CAS: 78-39-7 | C8H18O3 | 162.229 g/mol

Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl orthoacetate is used as pharmaceutical and water scavenger. It is also used in organic synthesis to introduce the acetate group into an alcohol. It is involved in the Johnson-Claisen rearrangement.

Solubility
Slightly miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 78-39-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD00009223
InChI Key NDQXKKFRNOPRDW-UHFFFAOYSA-N
Synonym triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
PubChem CID 66221
IUPAC Name 1,1,1-triethoxyethane
SMILES CCOC(C)(OCC)OCC

Specifications

Density 0.887
Boiling Point 142°C to 143°C
Flash Point 32°C (89°F)
Refractive Index 1.397
Quantity 500 mL
UN Number UN1993
Beilstein 506201
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material Triethyl orthoacetate
Show More Show Less
Hazard Category H226-H315-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 201-112-4
RTECSNumber KJ4075000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only