missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Citrate 99.0+%, TCI America™

Catalog No. C036725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C036725G 25 g
C0367500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C036725G Supplier TCI America Supplier No. C036725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.285
MDL Number MFCD00009201
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
Synonym triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
PubChem CID 6506
IUPAC Name triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O

Specifications

Melting Point -45°C
Color Yellow
Boiling Point 294°C
Quantity 25 g
Formula Weight 276.29
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Citrate
EINECSNumber (2)-1320
RTECSNumber GE8050000
TSCA Yes