missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl Borate 97.0+%, TCI America™
Supplier: TCI America B0520100ML
Specifications
| Triethyl Borate | |
| -85°C | |
| 118°C | |
| MFCD00009073 | |
| 1176 | |
| AJSTXXYNEIHPMD-UHFFFAOYSA-N | |
| triethyl borate | |
| 9009 | |
| 145.99 | |
| Liquid |
| 150-46-9 | |
| Yellow | |
| C6H15BO3 | |
| 100 mL | |
| triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester | |
| B(OCC)(OCC)OCC | |
| 145.993 | |
| CHEBI:38916 | |
| ≥97.0% (T) |
Chemical Identifiers
| 150-46-9 | |
| 145.993 | |
| AJSTXXYNEIHPMD-UHFFFAOYSA-N | |
| 9009 | |
| triethyl borate |
| C6H15BO3 | |
| MFCD00009073 | |
| triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester | |
| CHEBI:38916 | |
| B(OCC)(OCC)OCC |
Safety and Handling
EINECSNumber : (2)-2101
RTECSNumber : ED5075000
TSCA : Yes