missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyl 4-phosphonocrotonate, 92%, predominantly trans
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
Specifications
Specifications
| Color | Yellow |
| Density | 1.1200g/mL |
| Boiling Point | 133.0°C to 135.0°C (0.4 mmHg) |
| Flash Point | >112°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 92% |
| Packaging | Glass bottle |
| Refractive Index | 1.4525 to 1.4555 |
| Linear Formula | (C2H5O)2P(O)CH2CH=CHCO2C2H5 |
| Quantity | 10 g |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present |
RUO – Research Use Only