missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 4-phosphonocrotonate, 92%, predominantly trans

Catalog No. AC152120100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC152120100 10 g
AC152120500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152120100 Supplier Thermo Scientific Chemicals Supplier No. 152120100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10236-14-3 | C10H19O5P | 250.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 10236-14-3
Molecular Formula C10H19O5P
Molecular Weight (g/mol) 250.23
MDL Number MFCD00009192
InChI Key LXLODBXSCRTXFG-BQYQJAHWSA-N
Synonym triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
PubChem CID 5365783
SMILES CCOC(=O)\C=C\CP(=O)(OCC)OCC

Specifications

Color Yellow
Density 1.1200g/mL
Boiling Point 133.0°C to 135.0°C (0.4 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 92%
Packaging Glass bottle
Refractive Index 1.4525 to 1.4555
Linear Formula (C2H5O)2P(O)CH2CH=CHCO2C2H5
Quantity 10 g
Specific Gravity 1.12
IUPAC Name ethyl (E)-4-diethoxyphosphorylbut-2-enoate
Formula Weight 250.23
Percent Purity 92%
Physical Form Liquid
Chemical Name or Material Triethyl 4-phosphonocrotonate, 92%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present

RUO – Research Use Only