missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 4-phosphonobutyrate, 97%

Catalog No. p-7045376
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1048804 2 g
AAL1048809 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1048804 Supplier Thermo Scientific Chemicals Supplier No. L1048804
Only null left

CAS: 2327-69-7 | C10H21O5P | 252.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2327-69-7
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00015168
InChI Key FYESTJSRASJZOY-UHFFFAOYSA-N
PubChem CID 281201
IUPAC Name ethyl 4-diethoxyphosphorylbutanoate
SMILES CCOC(=O)CCCP(=O)(OCC)OCC

Specifications

Boiling Point 117°C to 119°C (0.4 mmHg)
Refractive Index 1.437
Quantity 2 g
Beilstein 1791802
Synonym triethyl 4-phosphonobutyrate, ethyl 4-diethoxyphosphoryl butanoate, triethyl4-phosphonobutyrate, butanoic acid, 4-diethoxyphosphinyl-, ethyl ester, triethyl-4-phosphono butyrate, butyric acid, 4-diethylphosphono-, ethyl ester, triethyl 4-phosphonobutanoate, 4-phosphonobutanoic acid triethyl ester, 4-diethoxyphosphinyl butyric acid ethyl ester, butanoic acid,4-diethoxyphosphinyl-,ethyl ester
Formula Weight 252.25
Percent Purity 97%
Chemical Name or Material Triethyl 4-phosphonobutyrate
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only