missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 3-Phosphonopropionate 98.0+%, TCI America™

Catalog No. p-7126766
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D152425G 25 g
D1524250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D152425G Supplier TCI America Supplier No. D152425G
Only null left

Chemical Identifiers

CAS 3699-67-0
Molecular Formula C9H19O5P
Molecular Weight (g/mol) 238.22
MDL Number MFCD00015167
InChI Key JMJWCUOIOKBVNQ-UHFFFAOYSA-N
Synonym ethyl 3-diethoxyphosphoryl propanoate, triethyl 3-phosphonopropionate, 3-diethylphosphonopropionic acid ethyl ester, triethyl 3-phosphonopropanoate, ethyl 3-diethoxyphosphinyl propionate, ethyl 3-diethoxycarbonyl propanoate, propanoic acid, 3-diethoxyphosphinyl-, ethyl ester, ethyl 3-diethylphosphonopropionate, ethyl 3-diethylphosphono-propionate, ethyl 3-diethylphosphono propanoate
PubChem CID 281204
IUPAC Name ethyl 3-diethoxyphosphorylpropanoate
SMILES CCOC(=O)CCP(=O)(OCC)OCC

Specifications

Color Colorless
Boiling Point 95°C
Quantity 25 g
Formula Weight 238.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl 3-Phosphonopropionate