missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonopropionate, 98%

Catalog No. p-7026925
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2326109 10 g
AAB2326118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2326109 Supplier Thermo Scientific Chemicals Supplier No. B2326109
Only null left

CAS: 3699-66-9 | C9H19O5P | 238.22 g/mol

Triethyl 2-phosphonopropionate is used as a reactant in intramolecular conjugate addition for the synthesis of floresolide B, enantioselective synthesis of spiroindane di-methyl acetic acid and in stereo selective intramolecular Diels-Alder reactions. It plays an important role in Horner-Wadsworth-Emmons reactions and chemoenzymatic one-pot synthesis of gamma-butyrolactones. It is also used in the preparation of 3-[2]furyl-2-methyl-acrylic acid ethyl ester by reacting with furfural.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 2-phosphonopropionate is used as a reactant in intramolecular conjugate addition for the synthesis of floresolide B, enantioselective synthesis of spiroindane di-methyl acetic acid and in stereo selective intramolecular Diels-Alder reactions. It plays an important role in Horner-Wadsworth-Emmons reactions and chemoenzymatic one-pot synthesis of gamma-butyrolactones. It is also used in the preparation of 3-[2]furyl-2-methyl-acrylic acid ethyl ester by reacting with furfural.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with strong bases.

Chemical Identifiers

CAS 3699-66-9
Molecular Formula C9H19O5P
Molecular Weight (g/mol) 238.22
MDL Number MFCD00009159
InChI Key BVSRWCMAJISCTD-UHFFFAOYSA-N
Synonym triethyl 2-phosphonopropionate, ethyl 2-diethoxyphosphoryl propanoate, ethyl-2-diethylphosphono propanoate, diethyl 1-carbethoxy ethylphosphonate, propanoic acid, 2-diethoxyphosphinyl-, ethyl ester, diethyl ethoxycarbonylethyl-phosphonate, diethyl 1-ethoxycarbonyl ethanephosphonate, 2-diethoxyphosphinyl propanoic acid ethyl ester, diethyl 1-ethoxycarbonyl ethanephosphonate∼2-phosphonopropionic acid triethyl ester, triethyl2-phosphonopropionate
PubChem CID 107155
IUPAC Name ethyl 2-diethoxyphosphorylpropanoate
SMILES CCOC(=O)C(C)P(=O)(OCC)OCC

Specifications

Density 1.111
Boiling Point 143°C to 144°C (12 mmHg)
Flash Point 88°C (190°F)
Odor Odorless
Refractive Index 1.432
Quantity 10 g
Beilstein 1786994
Solubility Information Miscible with chloroform and methanol.
Formula Weight 238.22
Percent Purity 98%
Chemical Name or Material Triethyl 2-phosphonopropionate
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 223-033-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only