missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonobutyrate, 98%, Thermo Scientific Chemicals

Catalog No. p-2692385
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC368780250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC368780250 Supplier Thermo Scientific Chemicals Supplier No. 368780250
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)

Chemical Identifiers

CAS 17145-91-4
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.25
MDL Number MFCD00041347
InChI Key GYUCVQSNZFRDRL-UHFFFAOYSA-N
Synonym triethyl 2-phosphonobutyrate, ethyl 2-diethoxyphosphoryl butanoate, ethyl 2-diethylphosphono butyrate, butanoic acid, 2-diethoxyphosphinyl-, ethyl ester, triethyl 2-phosphonobutanoate, .alpha.-diethylphosphono butanoic acid, ethyl ester, butyric acid, 2-phosphono-, triethyl ester, butyric acid, triethyl ester, triethyl 2-phosphonobu-tyrate, triethyl-2-phosphono butyrate
PubChem CID 229053
IUPAC Name ethyl 2-diethoxyphosphorylbutanoate
SMILES CCC(C(=O)OCC)P(=O)(OCC)OCC

Specifications

Color Colorless
Density 1.0640g/mL
Boiling Point 152.0°C to 154.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4315 to 1.4335
Linear Formula (O)P(CH3CH2O)2CH(CH2CH3)C(O)OCH2CH3
Quantity 25 mL
Specific Gravity 1.064
Formula Weight 252.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Triethyl 2-phosphonobutyrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only