missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-Fluoro-2-phosphonoacetate 95.0+%, TCI America™

Catalog No. p-7129623
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03401G 1 g
F03405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F03401G Supplier TCI America Supplier No. F03401G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2356-16-3
Molecular Formula C8H16FO5P
Molecular Weight (g/mol) 242.183
MDL Number MFCD00134400
InChI Key FVPISMANESAJQZ-UHFFFAOYSA-N
Synonym triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester
PubChem CID 2734865
IUPAC Name ethyl 2-diethoxyphosphoryl-2-fluoroacetate
SMILES CCOC(=O)C(F)P(=O)(OCC)OCC

Specifications

Color Yellow
Boiling Point 200°C
Quantity 1 g
Formula Weight 242.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl 2-Fluoro-2-phosphonoacetate