missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-fluoro-2-phosphonoacetate, 96%

CAS: 2356-16-3 | C8H16FO5P | 242.183 g/mol

Supplier:  Thermo Scientific Chemicals L1791306

 View more versions of this product

Catalog No. AAL1791306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Triethyl 2-fluoro-2-phosphonoacetate
1.194
73°C (163°F)
1.425
5 g
triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester
CCOC(=O)C(F)P(=O)(OCC)OCC
242.183
242.19
2356-16-3
75°C (0.01 mmHg)
C8H16FO5P
MFCD00134400
1912161
FVPISMANESAJQZ-UHFFFAOYSA-N
ethyl 2-diethoxyphosphoryl-2-fluoroacetate
2734865
96%

Chemical Identifiers

2356-16-3
242.183
FVPISMANESAJQZ-UHFFFAOYSA-N
2734865
CCOC(=O)C(F)P(=O)(OCC)OCC
C8H16FO5P
MFCD00134400
triethyl 2-fluoro-2-phosphonoacetate, 2-fluoro-2-phosphonoacetic acid triethyl ester, ethyl 2-diethoxyphosphoryl-2-fluoroacetate, triethyl phosphonofluoroacetate, ethyl diethoxyphosphoryl fluoro acetate, triethyl-2-fluoro-2-phosphonoacetate, acetic acid, diethoxyphosphinyl fluoro-, ethyl ester, 2-fluoro-2-phosphonoacetic acidtriethylester, diethyl ethoxycarbonyl fluoromethyl phosphonate, diethoxyphosphinyl fluoroacetic acid ethyl ester
ethyl 2-diethoxyphosphoryl-2-fluoroacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only